Chemical Identifiers
CAS96736-11-7
IUPAC Name(2R,3R,4S,5R,6R)-6-{[(3aS,7R,7aS)-7-hydroxy-4-oxo-1H,3aH,4H,5H,6H,7H,7aH-imidazo[4,5-c]pyridin-2-yl]amino}-5-[(3S)-3,6-diaminohexanamido]-4-hydroxy-2-(hydroxymethyl)oxan-3-yl carbamate; sulfuric acid
Molecular FormulaC19H36N8O12S
InChI KeyUDVWKDBCMFLRQW-TWRCRAKCSA-N
SMILESOS(O)(=O)=O.[H][C@]12NC(N[C@@H]3O[C@H](CO)[C@H](OC(N)=O)[C@@H](O)[C@H]3NC(=O)C[C@@H](N)CCCN)=N[C@]1([H])C(=O)NC[C@H]2O
View more