Thermo Scientific Chemicals

1,3-Dibromoacetone, 95%, Thermo Scientific Chemicals

Catalog number: H37325.06
5 g, Each
Thermo Scientific Chemicals

1,3-Dibromoacetone, 95%, Thermo Scientific Chemicals

Catalog number: H37325.06
5 g, Each
Quantity
Catalog number: H37325.06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
816-39-7
IUPAC Name
1,3-dibromopropan-2-one
Molecular Formula
C3H4Br2O
InChI Key
LQQKDSXCDXHLLF-UHFFFAOYSA-N
SMILES
BrCC(=O)CBr
Appearance (Color)
White to pale yellow or pale brown
Melting Point (clear melt)
24.5-30.5?C
Form
Crystals or powder or crystalline powder or fused/lumpy solid
Assay (GC)
≥94.0%

Description

1,3-Dibromoacetone is used as an antibiotic and antimicrobial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dibromoacetone is used as an antibiotic and antimicrobial agent.

Solubility
Soluble in acetone, chloroform, dichloromethane and methanol.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents and heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text