Thermo Scientific Chemicals

Poly(propylene glycol) bis(2-aminopropyl ether), Mn ca 230, Thermo Scientific Chemicals

Catalog number: H37543.K4
2.5 L, Each
Thermo Scientific Chemicals

Poly(propylene glycol) bis(2-aminopropyl ether), Mn ca 230, Thermo Scientific Chemicals

Catalog number: H37543.K4
2.5 L, Each
Quantity
Catalog number: H37543.K4
also known as H37543-K4
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9046-10-0
IUPAC Name
pyrrolidine-3,4-diol
Molecular Formula
C4H9NO2
InChI Key
JCZPOYAMKJFOLA-UHFFFAOYNA-N
SMILES
OC1CNCC1O
Appearance (Color)
Colorless to pale yellow
Form
Liquid
Color (APHA)
≤25 Hazen
Comment
Primary amines ≥97% of total amines
Comment
Total acetylatable groups 8.3-9.1 meq/g

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text