Palladium, 5% on activated carbon powder, Type 487, Thermo Scientific Chemicals
Palladium, 5% on activated carbon powder, Type 487, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Palladium, 5% on activated carbon powder, Type 487, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H37587.06
also known as H37587-06
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7440-05-3
IUPAC Namepalladium
Molecular FormulaPd
InChI KeyKDLHZDBZIXYQEI-UHFFFAOYSA-N
SMILES[Pd]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark grey to black
FormPowder
CommentSupport: Type 487 carbon powder
Elemental AnalysisPalladium Pd: ≥4.50 to ≤5.50%
Loss on Drying≤5.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only