May contain small amounts of colored product from manufacturing process
Form
Powder or crystalline powder
Appearance (Color)
White to yellow, pale brown or pale pink
Purity (DSC)
≥96.0%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.