Thermo Scientific Chemicals

(S)-(-)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol, 97+%, 99+% ee, Thermo Scientific Chemicals

Catalog number: H37716.06
5 g, Each
Thermo Scientific Chemicals

(S)-(-)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol, 97+%, 99+% ee, Thermo Scientific Chemicals

Catalog number: H37716.06
5 g, Each
Quantity
Catalog number: H37716.06
also known as H37716-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
65355-00-2
IUPAC Name
5H,5'H,6H,6'H,7H,7'H,8H,8'H-[1,1'-binaphthalene]-2,2'-diol
Molecular Formula
C20H22O2
InChI Key
UTXIFKBYNJRJPH-UHFFFAOYSA-N
SMILES
OC1=C(C2=C(CCCC2)C=C1)C1=C(O)C=CC2=C1CCCC2
Water Content (Karl Fischer Titration)
≤0.5%
Melting Point
160-168°C
Optical Rotation
-48° ± 1.5° (c=1 in chloroform)
Appearance (Color)
White to cream
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text