Platinum(IV) nitrate, solution, Pt 15% w/w, Thermo Scientific Chemicals
Platinum(IV) nitrate, solution, Pt 15% w/w, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum(IV) nitrate, solution, Pt 15% w/w, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number H37737.03
also known as H37737-03
Price (USD)/ Each
83.65
Online exclusive
92.90 
Save 9.25 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
83.65
Online exclusive
92.90 
Save 9.25 (10%)
Add to cart
Platinum(IV) nitrate, solution, Pt 15% w/w, Thermo Scientific Chemicals
Catalog numberH37737.03
Price (USD)/ Each
83.65
Online exclusive
92.90 
Save 9.25 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialPlatinum(IV) nitrate, solution
CAS10102-09-7
Health Hazard 1H290-H314-H332
Health Hazard 2GHS H Statement
H272-H314-H318-H290-H317
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
May cause an allergic skin reaction.
Health Hazard 3P234-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
View more
Platinum(IV) nitrate acts as a precursor in the production of platinum dioxide on support surface.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only