Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Quantity | 5000 g |
Melting Point | >450°C |
CAS | 68855-54-9 |
Molecular Formula | SiO{2} |
Synonym | Diatomaceous Earth;Filter aid, flux calcined |
Unit Size | Each |
Quantity | 1000 g |
Melting Point | >450°C |
CAS | 68855-54-9 |
Molecular Formula | SiO{2} |
Synonym | Diatomaceous Earth;Filter aid, flux calcined |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Quantity | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
H37758.0I also known as H37758-0I | Each | 5000 g | Special offer: 139.65 164.00 Save 24.35 (14%) Ends: 15-Oct-2024 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
H37758.0B also known as H37758-0B | Each | 1000 g | Special offer: 65.65 77.00 Save 11.35 (14%) Ends: 15-Oct-2024 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|