3-Bromo-1H-indazole, 97%, Thermo Scientific Chemicals
3-Bromo-1H-indazole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromo-1H-indazole, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number H37775.03
also known as H37775-03
Price (USD)/ Each
66.20
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
66.20
Add to cart
3-Bromo-1H-indazole, 97%, Thermo Scientific Chemicals
Catalog numberH37775.03
Price (USD)/ Each
66.20
-
Add to cart
Chemical Identifiers
CAS40598-94-5
IUPAC Name3-bromo-2H-indazole
Molecular FormulaC7H5BrN2
InChI KeyHTKXRTUKPXEALT-UHFFFAOYSA-N
SMILESBrC1=C2C=CC=CC2=NN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (GC)≥96.0%
FormPowder
Melting Point (clear melt)141.0-151.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only