Thermo Scientific Chemicals

Praseodymium(III) chloride, ultra dry, 99.95% (REO), Thermo Scientific Chemicals

Catalog number: H37815.14
25 g, Each
Thermo Scientific Chemicals

Praseodymium(III) chloride, ultra dry, 99.95% (REO), Thermo Scientific Chemicals

Catalog number: H37815.14
25 g, Each
Quantity
Catalog number: H37815.14
also known as H37815-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10361-79-2
Elemental analysis by ICP
Total elemental impurities: ≤500ppm
Elemental analysis by ICP
≥99.95% (PrCl₃)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text