1,1,1-Tris(diphenylphosphinomethyl)ethane is used in chemicals and pharmaceutical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,1,1-Tris(diphenylphosphinomethyl)ethane is used in chemicals and pharmaceutical research.
Solubility
Insoluble in water.
Notes
Air sensitive. Store away from oxidizing agents.
RUO – Research Use Only
General References:
- M.Di Vaira, C.A.Ghilardi; S.Midollini; L.Sacconi. cyclo-Triphosphorus (.delta.-P3) as a ligand in cobalt and nickel complexes with 1,1,1-tris(diphenylphosphinomethyl)ethane. Formation and structures. J. Am. Chem. Soc. 1978, 100, (8),2550-2551
- L.Sacconi; S.Midollini. Halogeno-complexes of cobalt(I) and nickel(I) with 1,1,1-tris(diphenylphosphinomethyl)ethane. J. Chem. Soc., Dalton Trans. 1972, (12),1213-1216