Thermo Scientific Chemicals

2,2'-Bithiophene-5-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Catalog number: H37989.06
5 g, Each
Thermo Scientific Chemicals

2,2'-Bithiophene-5-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Catalog number: H37989.06
5 g, Each
Quantity
Catalog number: H37989.06
also known as H37989-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
479719-88-5
IUPAC Name
2-{[2,2'-bithiophen]-5-yl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Formula
C14H17BO2S2
InChI Key
HPOQARMSOPOZMW-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=CC=C(S1)C1=CC=CS1
Form
Solid or Semi-solid or Fused solid or Liquid
Proton NMR
Conform to structure
Appearance (Color)
Colorless to dark blue to dark yellow or green and white to dark blue to dark yellow or green
Assay (GC)
≥97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text