2-(2,2,2-Trimethylacetamido)pyridine-3-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals
2-(2,2,2-Trimethylacetamido)pyridine-3-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(2,2,2-Trimethylacetamido)pyridine-3-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H50015.MD
also known as H50015-MD
Price (USD)/ Each
-
Quantity:
250 mg
Request bulk or custom format
Specifications
Chemical Name or Material2-(2,2,2-Trimethylacetamido)pyridine-3-boronic acid pinacol ester
CAS532391-30-3
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
2-(2,2,2-Trimethylacetamido)pyridine-3-boronic acid pinacol ester is used as an intermediate in organic synthesis. Further, it is used in agrochemicals, dyestuffs and in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2,2,2-Trimethylacetamido)pyridine-3-boronic acid pinacol ester is used as an intermediate in organic synthesis. Further, it is used in agrochemicals, dyestuffs and in pharmaceuticals.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only