4-Amino-3-pyridinemethanol, 95%, Thermo Scientific Chemicals
4-Amino-3-pyridinemethanol, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Amino-3-pyridinemethanol, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H50033.03
also known as H50033-03
Price (USD)/ Each
185.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
185.00
Add to cart
4-Amino-3-pyridinemethanol, 95%, Thermo Scientific Chemicals
Catalog numberH50033.03
Price (USD)/ Each
185.00
-
Add to cart
Chemical Identifiers
CAS138116-34-4
IUPAC Name(4-aminopyridin-3-yl)methanol
Molecular FormulaC6H8N2O
InChI KeyWNBVEYMTVDMSFZ-UHFFFAOYSA-N
SMILESNC1=CC=NC=C1CO
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>96.0%
FormCrystalline powder
Appearance (Color)White to cream
4-Amino-3-pyridinemethanol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-3-pyridinemethanol is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and heat.
RUO – Research Use Only