Thermo Scientific Chemicals

4-Amino-3-pyridinemethanol, 95%, Thermo Scientific Chemicals

Catalog number: H50033.06
5 g, Each
Thermo Scientific Chemicals

4-Amino-3-pyridinemethanol, 95%, Thermo Scientific Chemicals

Catalog number: H50033.06
5 g, Each
Quantity
Catalog number: H50033.06
also known as H50033-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
138116-34-4
IUPAC Name
(4-aminopyridin-3-yl)methanol
Molecular Formula
C6H8N2O
InChI Key
WNBVEYMTVDMSFZ-UHFFFAOYSA-N
SMILES
NC1=CC=NC=C1CO
Form
Crystalline powder
Assay (unspecified)
>96.0%
Appearance (Color)
White to cream

Description

4-Amino-3-pyridinemethanol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-3-pyridinemethanol is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text