Thermo Scientific Chemicals

6-Bromo-2-chloro-1,8-naphthyridine, 96%, Thermo Scientific Chemicals

Catalog number: H50043.03
1 g, Each
Thermo Scientific Chemicals

6-Bromo-2-chloro-1,8-naphthyridine, 96%, Thermo Scientific Chemicals

Catalog number: H50043.03
1 g, Each
Quantity
Catalog number: H50043.03
also known as H50043-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
902837-40-5
IUPAC Name
6-bromo-2-chloro-1,8-naphthyridine
Molecular Formula
C8H4BrClN2
InChI Key
KXSBJEOTLVRREX-UHFFFAOYSA-N
SMILES
ClC1=CC=C2C=C(Br)C=NC2=N1
Appearance (Color)
Yellow to brown
Form
Powder
Assay (HPLC)
>95.0%

Description

6-Bromo-2-chloro-1,8-naphthyridine, is used as raw material for synthesis of pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromo-2-chloro-1,8-naphthyridine, is used as raw material for synthesis of pharmaceutical intermediates.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text