Thermo Scientific Chemicals

2-Chloro-4-iodonicotinic acid, 98%, Thermo Scientific Chemicals

Catalog number: H50049.03
1 g, Each
Thermo Scientific Chemicals

2-Chloro-4-iodonicotinic acid, 98%, Thermo Scientific Chemicals

Catalog number: H50049.03
1 g, Each
Quantity
Catalog number: H50049.03
also known as H50049-03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
544671-78-5
Appearance (Color)
Pale yellow
Form
Fine powder
Assay (Silylated GC)
>97.5%

Description

2-Chloro-4-iodonicotinic acid is used as a pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-4-iodonicotinic acid is used as a pharmaceutical intermediates.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text