Thermo Scientific Chemicals

2-Isopropoxypyridine-3-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: H50089.03
1 g, Each
Thermo Scientific Chemicals

2-Isopropoxypyridine-3-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: H50089.03
1 g, Each
Quantity
Catalog number: H50089.03
also known as H50089-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
885278-10-4
IUPAC Name
2-(propan-2-yloxy)pyridine-3-carbaldehyde
Molecular Formula
C9H11NO2
InChI Key
GNTQOKGIVMJHQG-UHFFFAOYSA-N
SMILES
CC(C)OC1=C(C=O)C=CC=N1
Appearance (Color)
Pale yellow
Form
Liquid
Assay (HPLC)
>97.5%

Description

Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediate.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text