Thermo Scientific Chemicals

Benzo[b]thiophene-3-carboxylic acid, 96%, Thermo Scientific Chemicals

Catalog number: H50105.06
5 g, Each
Thermo Scientific Chemicals

Benzo[b]thiophene-3-carboxylic acid, 96%, Thermo Scientific Chemicals

Catalog number: H50105.06
5 g, Each
Quantity
Catalog number: H50105.06
also known as H50105-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5381-25-9
IUPAC Name
1-benzothiophene-3-carboxylic acid
Molecular Formula
C9H6O2S
InChI Key
DRBLTQNCQJXSNU-UHFFFAOYSA-N
SMILES
OC(=O)C1=CSC2=CC=CC=C12
Appearance (Color)
Light yellow
Form
Powder
Assay (HPLC)
>95.0%

Description

Benzo[b]thiophene-3-carboxylic acid is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo[b]thiophene-3-carboxylic acid is used as organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text