Thermo Scientific Chemicals

6-(Cyclopropylmethoxy)pyridine-3-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: H50139.MD
250 mg, Each
Thermo Scientific Chemicals

6-(Cyclopropylmethoxy)pyridine-3-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: H50139.MD
250 mg, Each
Quantity
Catalog number: H50139.MD
also known as H50139-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
947191-69-7
IUPAC Name
2-(cyclopropylmethoxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Formula
C15H22BNO3
InChI Key
QEQNYJUPYICBCR-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=CN=C(OCC2CC2)C=C1
Assay (HPLC)
96.0% min

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text