Ethyl 2-chloro-3-pyridylglyoxylate, 97%, Thermo Scientific Chemicals
Ethyl 2-chloro-3-pyridylglyoxylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 2-chloro-3-pyridylglyoxylate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H50153.03
also known as H50153-03
Price (USD)/ Each
189.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
189.00
Add to cart
Ethyl 2-chloro-3-pyridylglyoxylate, 97%, Thermo Scientific Chemicals
Catalog numberH50153.03
Price (USD)/ Each
189.00
-
Add to cart
Chemical Identifiers
CAS902837-56-3
IUPAC Nameethyl 2-(2-chloropyridin-3-yl)-2-oxoacetate
Molecular FormulaC9H8ClNO3
InChI KeyZTIMVJIGNBYGJG-UHFFFAOYSA-N
SMILESCCOC(=O)C(=O)C1=C(Cl)N=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>96.0%
FormLiquid
Appearance (Color)Reddish brown
Ethyl 2-chloro-3-pyridylglyoxylate is used as a pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-chloro-3-pyridylglyoxylate is used as a pharmaceutical intermediates.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only