Thermo Scientific Chemicals

1,3,5-Trimethyl-1H-pyrazole-4-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: H50225.03
1 g, Each
Thermo Scientific Chemicals

1,3,5-Trimethyl-1H-pyrazole-4-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: H50225.03
1 g, Each
Quantity
Catalog number: H50225.03
also known as H50225-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2644-93-1
IUPAC Name
1,3,5-trimethyl-1H-pyrazole-4-carbaldehyde
Molecular Formula
C7H10N2O
InChI Key
HMTUBXVXHHITGO-UHFFFAOYSA-N
SMILES
CN1N=C(C)C(C=O)=C1C
Appearance (Color)
Yellow
Form
Solid
Assay (GC)
≥97.5%

Description

1,3,5-Trimethyl-1H-pyrazole-4-carboxaldehyde is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Trimethyl-1H-pyrazole-4-carboxaldehyde is used as organic chemical synthesis intermediate.

Notes
Air sensitive. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text