Thermo Scientific Chemicals

2-Cyanobenzoic acid, 94%, Thermo Scientific Chemicals

Catalog number: H50237.06
5 g, Each
Thermo Scientific Chemicals

2-Cyanobenzoic acid, 94%, Thermo Scientific Chemicals

Catalog number: H50237.06
5 g, Each
Quantity
Catalog number: H50237.06
also known as H50237-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3839-22-3
IUPAC Name
2-cyanobenzoate
Molecular Formula
C8H4NO2
InChI Key
DTNSDCJFTHMDAK-UHFFFAOYSA-M
SMILES
[O-]C(=O)C1=CC=CC=C1C#N
Appearance (Color)
White to pale cream or pale grey
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥92.0%

Description

2-Cyanobenzoic acid has been used as reactant in the synthesis of T-box riboswitch antiterminator RNA-binding oxazolidinone derivatives, Benzoate modified β-cyclodextrin derivatives and Pyrrolo[1,2-f]triazines for use as JAK2 inhibitors. As a reactant involved in decarboxylation, decarboxylative halogenation and protodecarboxylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyanobenzoic acid has been used as reactant in the synthesis of T-box riboswitch antiterminator RNA-binding oxazolidinone derivatives, Benzoate modified β-cyclodextrin derivatives and Pyrrolo[1,2-f]triazines for use as JAK2 inhibitors. As a reactant involved in decarboxylation, decarboxylative halogenation and protodecarboxylation.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text