Specifications
Chemical Name or Material1-Bromo-2-(bromomethyl)naphthalene
CAS37763-43-2
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more