Thermo Scientific Chemicals

Glycine isopropyl ester hydrochloride, 96%, Thermo Scientific Chemicals

Catalog number: H50249.06
5 g, Each
Thermo Scientific Chemicals

Glycine isopropyl ester hydrochloride, 96%, Thermo Scientific Chemicals

Catalog number: H50249.06
5 g, Each
Quantity
Catalog number: H50249.06
also known as H50249-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14019-62-6
IUPAC Name
propan-2-yl 2-aminoacetate hydrochloride
Molecular Formula
C5H12ClNO2
InChI Key
UBKCIXXGQRZHRO-UHFFFAOYSA-N
SMILES
Cl.CC(C)OC(=O)CN
Form
Powder
Appearance (Color)
White
Melting Point
97-104°C
Proton NMR
Conforms
Infrared spectrum
Conforms

Description

Glycine isopropyl ester hydrochloride can react with indol-3-yl-oxo-acetyl chloride to produce [2-(1H-indol-3-yl)-2-oxo-acetylamino]-acetic acid isopropyl ester. This reaction will need reagent Et3N, and the menstruum benzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine isopropyl ester hydrochloride can react with indol-3-yl-oxo-acetyl chloride to produce [2-(1H-indol-3-yl)-2-oxo-acetylamino]-acetic acid isopropyl ester. This reaction will need reagent Et3N, and the menstruum benzene.

Solubility
Soluble in water

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text