Thermo Scientific Chemicals

N,N-Dimethyl-1,4-butanediamine, 98%, Thermo Scientific Chemicals

Catalog number: H50276.06
5 g, Each
Thermo Scientific Chemicals

N,N-Dimethyl-1,4-butanediamine, 98%, Thermo Scientific Chemicals

Catalog number: H50276.06
5 g, Each
Quantity
Catalog number: H50276.06
also known as H50276-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3529-10-0
IUPAC Name
(4-aminobutyl)dimethylamine
Molecular Formula
C6H16N2
InChI Key
GCOWZPRIMFGIDQ-UHFFFAOYSA-N
SMILES
CN(C)CCCCN
Form
Liquid
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (GC)
≥97.5%
Refractive Index
1.4375-1.4415 @ 20?C
Appearance (Color)
Clear colorless

Description

N,N-Dimethyl-1,4-butanediamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyl-1,4-butanediamine is used as pharmaceutical intermediate.

Notes
Air sensitive. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text