Thermo Scientific Chemicals

N,N-Diethyl-1,4-butanediamine, 96%, Thermo Scientific Chemicals

Catalog number: H50278.03
1 g, Each
Thermo Scientific Chemicals

N,N-Diethyl-1,4-butanediamine, 96%, Thermo Scientific Chemicals

Catalog number: H50278.03
1 g, Each
Quantity
Catalog number: H50278.03
also known as H50278-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
27431-62-5
IUPAC Name
(4-aminobutyl)diethylamine
Molecular Formula
C8H20N2
InChI Key
JILXUIANNUALRZ-UHFFFAOYSA-N
SMILES
CCN(CC)CCCCN
Form
Liquid
Assay by NMR
>95.0%
Appearance (Color)
Colorless

Description

N,N-Diethyl-1,4-butanediamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Diethyl-1,4-butanediamine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text