4'-Fluoro-2-biphenylmethylamine, 97%, Thermo Scientific Chemicals
4'-Fluoro-2-biphenylmethylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4'-Fluoro-2-biphenylmethylamine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H50297.MD
also known as H50297-MD
Price (USD)/ Each
70.60
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
70.60
Add to cart
4'-Fluoro-2-biphenylmethylamine, 97%, Thermo Scientific Chemicals
Catalog numberH50297.MD
Price (USD)/ Each
70.60
-
Add to cart
Chemical Identifiers
CAS884504-18-1
IUPAC Name1-{4'-fluoro-[1,1'-biphenyl]-2-yl}methanamine
Molecular FormulaC13H12FN
InChI KeyMLXCERLVQPYBIA-UHFFFAOYSA-N
SMILESNCC1=CC=CC=C1C1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow
Assay (HPLC)>96.0%
FormOil
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only