1-(4-Pyridyl)ethylamine is used as a pharmaceutical intermediates
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-(4-Pyridyl)ethylamine is used as a pharmaceutical intermediates
Solubility
Soluble in water
Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
- Arthur J. Ashe. Trimethylsilyl migrations in the 5-trimethylsilylbicyclo[2.1.0]pentanes. J. Am. Chem. Soc. 1973, 95, (3), 818-821.
- Hiroshi. Ichikawa.; Michihiko. Ogata. Molecular orbital approach to the interpretation of organic mass spectra. III. Determination of intermediate ion structure in the mass spectrometry of heteroaromatic compounds. J. Am. Chem. Soc. 1973, 95, (3), 806-811.