Thermo Scientific Chemicals

4-(1-Pyrrolidinyl)-1-butylamine, 98%, Thermo Scientific Chemicals

Catalog number: H50323.03
1 g, Each
Thermo Scientific Chemicals

4-(1-Pyrrolidinyl)-1-butylamine, 98%, Thermo Scientific Chemicals

Catalog number: H50323.03
1 g, Each
Quantity
Catalog number: H50323.03
also known as H50323-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
24715-90-0
IUPAC Name
4-(pyrrolidin-1-yl)butan-1-amine
Molecular Formula
C8H18N2
InChI Key
LSDYCEIPEBJKPT-UHFFFAOYSA-N
SMILES
NCCCCN1CCCC1
Appearance (Color)
Clear to pale yellow
Form
Liquid
Assay (unspecified)
>97.5%

Description

4-(1-Pyrrolidinyl)-1-butylamine is used directly as drugs, food, cosmetics. Used as a building block in organic synthesis, pyrrolidine activates ketones and aldehydes toward nucleophilic addition by formation of enamines

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(1-Pyrrolidinyl)-1-butylamine is used directly as drugs, food, cosmetics. Used as a building block in organic synthesis, pyrrolidine activates ketones and aldehydes toward nucleophilic addition by formation of enamines

Solubility
Soluble in water

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text