4-Methoxy-1-butanol is used as a chemicals, pharmaceutical intermediates, reagents. 4-Methoxy-1-butanol is used with n-butyl acetate to improve dissolving power, drying time, and flow.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methoxy-1-butanol is used as a chemicals, pharmaceutical intermediates, reagents. 4-Methoxy-1-butanol is used with n-butyl acetate to improve dissolving power, drying time, and flow.
Solubility
Slightly soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Herbert C. Brown.; M. K. Unni. Hydroboration. XXV. The hydroboration of 3-butenyl derivatives containing representative substituents. J. Am. Chem. Soc. 1968, 90, (11), 2902-2905.
Herbert C. Brown.; Gary J. Lynch. Solvomercuration-demercuration. 8. Oxymercuration-demercuration of methoxy-, hydroxy-. and acetoxy-substituted alkenes. J. Org. Chem. 1981, 46, (3), 531-538.