Thermo Scientific Chemicals

2-n-Propyl-1-heptanol, 98%, Thermo Scientific Chemicals

Catalog number: H50340.03
1 g, Each
Thermo Scientific Chemicals

2-n-Propyl-1-heptanol, 98%, Thermo Scientific Chemicals

Catalog number: H50340.03
1 g, Each
Quantity
Catalog number: H50340.03
also known as H50340-03
Price (USD)
Quantity
-
Have Questions?

Description

2-n-Propyl-1-heptanol is used as a starting material for the production of plasticizers for PVC, e.g. Palatinol™ 10-P.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-n-Propyl-1-heptanol is used as a starting material for the production of plasticizers for PVC, e.g. Palatinol™ 10-P.

Solubility
Insoluble in water. Miscible with most common organic solvents.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text