Thermo Scientific Chemicals

3-Methoxy-4-methylaniline, 99+%, Thermo Scientific Chemicals

Catalog number: H50362.03
1 g, Each
Thermo Scientific Chemicals

3-Methoxy-4-methylaniline, 99+%, Thermo Scientific Chemicals

Catalog number: H50362.03
1 g, Each
Quantity
Catalog number: H50362.03
also known as H50362-03
Price (USD)
Quantity
-
Have Questions?

Description

3-Methoxy-4-methylaniline, afforded by an electrophilic substitution, using the 2-methoxy-substituted iron complex sail followed by oxidative cyclization with concomitant aromation of the resulting iron complex. It is used to produce 8-methoxy-7-methylalloxazine. This reaction will need solvent ethanol, H2O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxy-4-methylaniline, afforded by an electrophilic substitution, using the 2-methoxy-substituted iron complex sail followed by oxidative cyclization with concomitant aromation of the resulting iron complex. It is used to produce 8-methoxy-7-methylalloxazine. This reaction will need solvent ethanol, H2O

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text