3-Methoxy-4-methylaniline, afforded by an electrophilic substitution, using the 2-methoxy-substituted iron complex sail followed by oxidative cyclization with concomitant aromation of the resulting iron complex. It is used to produce 8-methoxy-7-methylalloxazine. This reaction will need solvent ethanol, H2O
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Methoxy-4-methylaniline, afforded by an electrophilic substitution, using the 2-methoxy-substituted iron complex sail followed by oxidative cyclization with concomitant aromation of the resulting iron complex. It is used to produce 8-methoxy-7-methylalloxazine. This reaction will need solvent ethanol, H2O
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
- Subhash C. Basak.; Gregory D. Grunwald. Molecular Similarity and Estimation of Molecular Properties. J. Chem. Inf. Comput. Sci. 1995, 35, (3), 366-372.
- Herbert C. Brown.; Gary J. Lynch. Prediction of heteroaromatic amine mutagenicity by means of correlation weighting of atomic orbital graphs of local invariants. Journal of Molecular Structure: THEOCHEM. 2001, 538, (1-3), 287-293.