Thermo Scientific Chemicals

4-Amino-N,N-dimethylbenzamide, 97+%, Thermo Scientific Chemicals

Catalog number: H50364.06
5 g, Each
Thermo Scientific Chemicals

4-Amino-N,N-dimethylbenzamide, 97+%, Thermo Scientific Chemicals

Catalog number: H50364.06
5 g, Each
Quantity
Catalog number: H50364.06
also known as H50364-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6331-71-1
IUPAC Name
4-amino-N,N-dimethylbenzamide
Molecular Formula
C9H12N2O
InChI Key
QEPGWLBMAAEBCP-UHFFFAOYSA-N
SMILES
CN(C)C(=O)C1=CC=C(N)C=C1
Form
Powder
Assay (HPLC)
≥97%
Proton NMR
Conforms
Appearance (Color)
Pale grey

Description

4-Amino-N,N-dimethylbenzamide is an important raw material and intermediate used in pharmaceuticals, agro based and dye stuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-N,N-dimethylbenzamide is an important raw material and intermediate used in pharmaceuticals, agro based and dye stuff fields.

Solubility
Very soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text