3-Amino-1-bromoisoquinoline, 97+%, Thermo Scientific Chemicals
3-Amino-1-bromoisoquinoline, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Amino-1-bromoisoquinoline, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H50368.03
also known as H50368-03
Price (USD)/ Each
211.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
211.00
Add to cart
3-Amino-1-bromoisoquinoline, 97+%, Thermo Scientific Chemicals
Catalog numberH50368.03
Price (USD)/ Each
211.00
-
Add to cart
Chemical Identifiers
CAS13130-79-5
IUPAC Name1-bromoisoquinolin-3-amine
Molecular FormulaC9H7BrN2
InChI KeyPSQUIUNIVDKHJK-UHFFFAOYSA-N
SMILESNC1=NC(Br)=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)97+%
3-Amino-1-bromoisoquinoline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-1-bromoisoquinoline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Francis Johnson; W.A.Nasutavicus. Polyfunctional aliphatic compounds. IV. The cyclization of nitriles by halogen acids. A new synthesis of thiazoles. J. Org. Chem. 1963, 28, (7),1877-1883
  2. Hannah Weiler-Feilchenfeld, Yuh-Lin Mao; E.D.Bergmann. The dipole moments of some halogen substituted isoquinolines. Israel Journal of Chemistry. 1971, 9, (2),111-118