Thermo Scientific Chemicals

2-(Phthalimido)ethanesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: H50382.03
1 g, Each
Thermo Scientific Chemicals

2-(Phthalimido)ethanesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: H50382.03
1 g, Each
Quantity
Catalog number: H50382.03
also known as H50382-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4403-36-5
IUPAC Name
2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)ethane-1-sulfonyl chloride
Molecular Formula
C10H8ClNO4S
InChI Key
HCPVYBCAYPMANM-UHFFFAOYSA-N
SMILES
ClS(=O)(=O)CCN1C(=O)C2=CC=CC=C2C1=O
Form
Powder
Assay (HPLC)
≥96.0%
Appearance (Color)
White to cream
Melting Point (clear melt)
155-163?C
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%

Description

2-(Phthalimido)ethanesulfonyl chloride is an important raw material and intermediate used in pharmaceuticals, agro based and dye stuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Phthalimido)ethanesulfonyl chloride is an important raw material and intermediate used in pharmaceuticals, agro based and dye stuff fields.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text