3-(4-Chlorosulfonylphenyl)propionic acid is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-(4-Chlorosulfonylphenyl)propionic acid is used as pharmaceutical intermediate.
Solubility
It reacts with water.
Notes
Moisture sensitive. Incompatible with oxidizing agents, bases and active metals.
RUO – Research Use Only
Elisabetta Maria Usai; Maria Franca Sini; Daniela Meloni, Vincenzo Solinas; Andrea Salis. Sulfonic acid-functionalized mesoporous silicas: Microcalorimetric characterization and catalytic performance toward biodiesel synthesis. Microporous and Mesoporous Materials. 2013, 179, 54-62