1-Naphthylacetyl chloride, 95%, Thermo Scientific Chemicals
1-Naphthylacetyl chloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Naphthylacetyl chloride, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H50386.06
also known as H50386-06
Price (USD)/ Each
480.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
480.00
Add to cart
1-Naphthylacetyl chloride, 95%, Thermo Scientific Chemicals
Catalog numberH50386.06
Price (USD)/ Each
480.00
-
Add to cart
Chemical Identifiers
CAS5121-00-6
IUPAC Name2-(naphthalen-1-yl)acetyl chloride
Molecular FormulaC12H9ClO
InChI KeyDSVAZLXLRDXHKO-UHFFFAOYSA-N
SMILESClC(=O)CC1=CC=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormLiquid
Assay (unspecified)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only