3-(4-Methoxyphenyl)propionyl chloride is an important raw material and intermediate used in pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-(4-Methoxyphenyl)propionyl chloride is an important raw material and intermediate used in pharmaceuticals, agrochemicals and dyestuff.
Solubility
Reacts with water.
Notes
Air & Moisture Sensitive. Protect from humidity and water. Store under dry inert gas away from oxidizing agents and moisture. Incompatible with bases.
RUO – Research Use Only