5-Bromo-2-fluoro-m-xylene, 97%, Thermo Scientific Chemicals
5-Bromo-2-fluoro-m-xylene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Bromo-2-fluoro-m-xylene, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H50564.03
also known as H50564-03
Price (USD)/ Each
49.30
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
49.30
Add to cart
5-Bromo-2-fluoro-m-xylene, 97%, Thermo Scientific Chemicals
Catalog numberH50564.03
Price (USD)/ Each
49.30
-
Add to cart
Chemical Identifiers
CAS99725-44-7
IUPAC Name5-bromo-2-fluoro-1,3-dimethylbenzene
Molecular FormulaC8H8BrF
InChI KeyZXPHUVHMBKRRJF-UHFFFAOYSA-N
SMILESCC1=CC(Br)=CC(C)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)96.0% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only