7-Bromo-1-tetralone, 97%, Thermo Scientific Chemicals
7-Bromo-1-tetralone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

7-Bromo-1-tetralone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H50604.06
also known as H50604-06
Price (USD)/ Each
524.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
524.00
Add to cart
7-Bromo-1-tetralone, 97%, Thermo Scientific Chemicals
Catalog numberH50604.06
Price (USD)/ Each
524.00
-
Add to cart
Chemical Identifiers
CAS32281-97-3
IUPAC Name7-bromo-1,2,3,4-tetrahydronaphthalen-1-one
Molecular FormulaC10H9BrO
InChI KeyYGVDCGFUUUJCDF-UHFFFAOYSA-N
SMILESBrC1=CC=C2CCCC(=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to brown
Assay (GC)≥96.0%
Melting Point (clear melt)71.0-80.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only