2-Bromo-5-methoxybenzonitrile, 98%, Thermo Scientific Chemicals
2-Bromo-5-methoxybenzonitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-5-methoxybenzonitrile, 98%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
250 mg
Catalog number H50608.MD
also known as H50608-MD
Price (USD)/ Each
65.50
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
65.50
Add to cart
2-Bromo-5-methoxybenzonitrile, 98%, Thermo Scientific Chemicals
Catalog numberH50608.MD
Price (USD)/ Each
65.50
-
Add to cart
Chemical Identifiers
CAS138642-47-4
IUPAC Name2-bromo-5-methoxybenzonitrile
Molecular FormulaC8H6BrNO
InChI KeyZQONVYONOASKIY-UHFFFAOYSA-N
SMILESCOC1=CC(C#N)=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)97.5% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
 
RUO – Research Use Only