Thermo Scientific Chemicals

Dimethyl 5-bromoisophthalate, 98%, Thermo Scientific Chemicals

Catalog number: H50629.03
1 g, Each
Thermo Scientific Chemicals

Dimethyl 5-bromoisophthalate, 98%, Thermo Scientific Chemicals

Catalog number: H50629.03
1 g, Each
Quantity
Catalog number: H50629.03
also known as H50629-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
51760-21-5
IUPAC Name
1,3-dimethyl 5-bromobenzene-1,3-dicarboxylate
Molecular Formula
C10H9BrO4
InChI Key
QUJINGKSNJNXEB-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC(=CC(Br)=C1)C(=O)OC
Assay (GC)
97.5% min

Description

Dimethyl 5-bromoisophthalate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl 5-bromoisophthalate is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text