Thermo Scientific Chemicals

Ethyl 3-tert-butyl-1H-pyrazole-5-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: H50678.03
1 g, Each
Thermo Scientific Chemicals

Ethyl 3-tert-butyl-1H-pyrazole-5-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: H50678.03
1 g, Each
Quantity
Catalog number: H50678.03
also known as H50678-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
916791-97-4
IUPAC Name
ethyl 5-tert-butyl-1H-pyrazole-3-carboxylate
Molecular Formula
C10H16N2O2
InChI Key
RCXMILPYEKVQLB-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=NNC(=C1)C(C)(C)C
Assay (HPLC)
96.0% min

Description

Ethyl 3-tert-butyl-1H-pyrazole-5-carboxylate is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-tert-butyl-1H-pyrazole-5-carboxylate is used as pharmaceutical intermediates.

Notes
Store in a cool, dry condition in a well sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text