2'-Hydroxy-4',5'-dimethylacetophenone, 98%, Thermo Scientific Chemicals
2'-Hydroxy-4',5'-dimethylacetophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2'-Hydroxy-4',5'-dimethylacetophenone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H50685.03
also known as H50685-03
Price (USD)/ Each
49.50
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
49.50
Add to cart
2'-Hydroxy-4',5'-dimethylacetophenone, 98%, Thermo Scientific Chemicals
Catalog numberH50685.03
Price (USD)/ Each
49.50
-
Add to cart
Chemical Identifiers
CAS36436-65-4
IUPAC Name1-(2-hydroxy-4,5-dimethylphenyl)ethan-1-one
Molecular FormulaC10H12O2
InChI KeyYXVSURZEXVMUAM-UHFFFAOYSA-N
SMILESCC(=O)C1=CC(C)=C(C)C=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream
FormPowder
Proton NMRConforms
Melting Point68-70?C
2'-Hydroxy-4',5'-dimethylacetophenone is used in the preparation of chalcone derivatives with antibacterial activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Hydroxy-4′,5′-dimethylacetophenone is used in the preparation of chalcone derivatives with antibacterial activity.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry condition in a well sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only
Zheng Lu; Corbet White; Arnold L. Rheingold; Robert H. Crabtree. Deprotonated thioamides as thiolate S-donor ligands with a high tendency to avoid M-S-M bridge formation: crystal and molecular structure of bis(2-hydroxy-5-methylacetophenone N,N-dimethylthiosemicarbazonato)dinickel. Inorg. Chem.1993, 32, (19), 3991-3994.