Thermo Scientific Chemicals

5-Amino-2-(4-morpholinyl)pyridine, 97%, Thermo Scientific Chemicals

Catalog number: H50699.06
5 g, Each
Thermo Scientific Chemicals

5-Amino-2-(4-morpholinyl)pyridine, 97%, Thermo Scientific Chemicals

Catalog number: H50699.06
5 g, Each
Quantity
Catalog number: H50699.06
also known as H50699-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
52023-68-4
IUPAC Name
5-amino-2-(morpholin-4-yl)pyridin-1-ium
Molecular Formula
C9H14N3O
InChI Key
VVTSPTCBHTWXMD-UHFFFAOYSA-O
SMILES
NC1=CC=C([NH+]=C1)N1CCOCC1
Appearance (Color)
Grey to dark purple
Form
Solid
Assay (HPLC)
>96.0%

Description

5-Amino-2-(4-morpholinyl)pyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Amino-2-(4-morpholinyl)pyridine is used as pharmaceutical intermediate.

Notes
Store at room temperature. Store in a cool, dry condition in a well sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text