Piperonylic acid hydrazide, 97%, Thermo Scientific Chemicals
Piperonylic acid hydrazide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Piperonylic acid hydrazide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H50713.03
also known as H50713-03
Price (USD)/ Each
44.70
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
44.70
Add to cart
Piperonylic acid hydrazide, 97%, Thermo Scientific Chemicals
Catalog numberH50713.03
Price (USD)/ Each
44.70
-
Add to cart
Specifications
Chemical Name or MaterialPiperonylic acid hydrazide
CAS22026-39-7
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Piperonylic acid hydrazide is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Piperonylic acid hydrazide is used as organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Kenneth Hutton. The synthesis of some new phenylurethans as potential local anesthetics. J. Org. Chem. 1955, 20, (7),855-861
  2. M.P.Cava; M.Srinivasan. The synthesis of N-methylovigerine. Tetrahedron. 1970, 26, (19),4649-4653