Thermo Scientific Chemicals

2-Phenoxypropylamine, 98%, Thermo Scientific Chemicals

Catalog number: H50720.MD
250 mg, Each
Thermo Scientific Chemicals

2-Phenoxypropylamine, 98%, Thermo Scientific Chemicals

Catalog number: H50720.MD
250 mg, Each
Quantity
Catalog number: H50720.MD
also known as H50720-MD
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
6437-49-6
IUPAC Name
2-phenoxypropan-1-amine hydrochloride
Molecular Formula
C9H14ClNO
InChI Key
CGOBBGUREQJPPH-UHFFFAOYNA-N
SMILES
Cl.CC(CN)OC1=CC=CC=C1
Assay (HPLC)
97.5% min

Description

2-Phenoxypropylamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Phenoxypropylamine is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 1
    Serious Eye Damage/Eye Irritation
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H314 - Causes severe skin burns and eye damage

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P330 + P331 - IF SWALLOWED: Rinse mouth. Do NOT induce vomiting

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician