2-Methyl-2-(4-methylphenyl)morpholine, 97%, Thermo Scientific Chemicals
2-Methyl-2-(4-methylphenyl)morpholine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methyl-2-(4-methylphenyl)morpholine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H50732.MD
also known as H50732-MD
Price (USD)/ Each
87.40
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
87.40
Add to cart
2-Methyl-2-(4-methylphenyl)morpholine, 97%, Thermo Scientific Chemicals
Catalog numberH50732.MD
Price (USD)/ Each
87.40
-
Add to cart
Chemical Identifiers
CAS902836-81-1
IUPAC Name2-methyl-2-(4-methylphenyl)morpholine
Molecular FormulaC12H17NO
InChI KeyNQYMBSUCKZVKFF-UHFFFAOYNA-N
SMILESCC1=CC=C(C=C1)C1(C)CNCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)97%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only